Learn More
ILEX LIFE SCIENCES LLC PAN Biotech CMRL 1066 Cell Culture Medium without L Glutamine without Phenol Red GMP Grade 500 ML 1/EA
SDP
Catalog No. 5000580290
Encompass_Preferred
PAN-Biotech CMRL 1066 Cell Culture Medium w/o L-Glutamine w/o Phenol red w 2 2 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION CMRL-1066 GMP Grade is a ready-to-use chemically-defined medium designed for direct use in GMP-regulated clinical and industrial applications It provides a precisely controlled formulation that supports stable and reproducible cell culture performance Combined with high batch-to-batch consistency controlled quality parameters and comprehensive documentation CMRL-1066 GMP is a true ready-to-use medium suitable for sensitive development scale-up and manufacturing workflows QUALITY GMP Grade (EU Annex 1) BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE No Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED Without Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80290 Supplier ILEX LIFE SCIENCES LLC Supplier No. CT84600
PAN-Biotech CMRL 1066 Cell Culture Medium w/o L-Glutamine w/o Phenol red w 2 2 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION CMRL-1066 GMP Grade is a ready-to-use chemically-defined medium designed for direct use in GMP-regulated clinical and industrial applications It provides a precisely controlled formulation that supports stable and reproducible cell culture performance Combined with high batch-to-batch consistency controlled quality parameters and comprehensive documentation CMRL-1066 GMP is a true ready-to-use medium suitable for sensitive development scale-up and manufacturing workflows QUALITY GMP Grade (EU Annex 1) BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE No Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED Without Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany