Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Cryopan III Freezing Medium Chemically Defined serum and DMSO free 500 ML 1/EA
SDP
Catalog No. 5000580277
Encompass_Preferred
PAN-Biotech Cryopan III Chemically-Defined Serum-Free DMSO-Free Freezing Medium (500 ml) Cryopreservation Media DESCRIPTION PAN-Biotech Cryopan III is a chemically-defined serum-free freezing medium without DMSO that is suitable for the cryopreservation of both primary cells and established cell lines Its chemically defined composition free from human or animal components ensures optimal cell preservation Cryopan III can be used for freezing adherent or suspension cells making it a versatile option for the cryopreservation of animal and human cells across different cell types and culture conditions The medium has also been proven effective for preserving tissue samples e g umbilical cord tissue The formulation without DMSO is particularly well-suited for primary cells and other applications in which the cytotoxic side-effects associated with DMSO are not desired
Catalog No. 50-005-80277 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0796500
PAN-Biotech Cryopan III Chemically-Defined Serum-Free DMSO-Free Freezing Medium (500 ml) Cryopreservation Media DESCRIPTION PAN-Biotech Cryopan III is a chemically-defined serum-free freezing medium without DMSO that is suitable for the cryopreservation of both primary cells and established cell lines Its chemically defined composition free from human or animal components ensures optimal cell preservation Cryopan III can be used for freezing adherent or suspension cells making it a versatile option for the cryopreservation of animal and human cells across different cell types and culture conditions The medium has also been proven effective for preserving tissue samples e g umbilical cord tissue The formulation without DMSO is particularly well-suited for primary cells and other applications in which the cytotoxic side-effects associated with DMSO are not desired