Learn More
ILEX LIFE SCIENCES LLC PAN Biotech DMEM Cell Culture Medium High Glucose With Stable Glutamine Without Sodium Pyruvate GMP Grade 500 ML 1/EA
SDP
Catalog No. 5000580393
Encompass_Preferred
PAN-Biotech DMEM Cell Culture Medium w 4 5 g/L Glucose w stable Glutamine w/o Sodium pyruvate w 3 7 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION PAN-Biotech DMEM GMP Grade is a ready-to-use classical basal medium designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the well-established DMEM formulation and is manufactured under strictly controlled GMP conditions Defined raw materials high batch-to-batch consistency and comprehensive documentation make DMEM GMP a true ready-to-use solution suitable for immediate use in development scale-up and manufacturing workflows QUALITY GMP Grade (EU Annex 1) BICARBONATE With 3 7 g/L Sodium Bicarbonate GLUCOSE High Glucose (4 5 g/L) GLUTAMINE With Stable Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80393 Supplier ILEX LIFE SCIENCES LLC Supplier No. CT04500
PAN-Biotech DMEM Cell Culture Medium w 4 5 g/L Glucose w stable Glutamine w/o Sodium pyruvate w 3 7 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION PAN-Biotech DMEM GMP Grade is a ready-to-use classical basal medium designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the well-established DMEM formulation and is manufactured under strictly controlled GMP conditions Defined raw materials high batch-to-batch consistency and comprehensive documentation make DMEM GMP a true ready-to-use solution suitable for immediate use in development scale-up and manufacturing workflows QUALITY GMP Grade (EU Annex 1) BICARBONATE With 3 7 g/L Sodium Bicarbonate GLUCOSE High Glucose (4 5 g/L) GLUTAMINE With Stable Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany