Learn More
ILEX LIFE SCIENCES LLC PAN Biotech DMEM Cell Culture Medium Low Glucose with Stable Glutamine with Sodium Pyruvate 500 ML 1/EA
SDP
Catalog No. 5000580432
Encompass_Preferred
PAN-Biotech DMEM Cell Culture Medium w 1 0 g/L Glucose w stable Glutamine w Sodium pyruvate w 2 2 g/L NaHCO3 500 ml INTRODUCTION Intrinsically developed for the cultivation of murine embryonic cells DMEM (Dulbecco s Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells especially if the medium is supplemented with FBS DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE Low Glucose (1 0 g/L) GLUTAMINE With Stable Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80432 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0401504
PAN-Biotech DMEM Cell Culture Medium w 1 0 g/L Glucose w stable Glutamine w Sodium pyruvate w 2 2 g/L NaHCO3 500 ml INTRODUCTION Intrinsically developed for the cultivation of murine embryonic cells DMEM (Dulbecco s Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells especially if the medium is supplemented with FBS DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE Low Glucose (1 0 g/L) GLUTAMINE With Stable Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures