Learn More
ILEX LIFE SCIENCES LLC PAN Biotech DMSO Dimethyl Sulfoxide Solution for cell culture endotoxin tested 100 ML 1/EA
SDP
Catalog No. 5000580173
Encompass_Preferred
PAN-Biotech Dimethyl Sulfoxide (DMSO) for Cell Culture Endotoxin Tested 100 ml DESCRIPTION PAN-Biotech Dimethyl Sulfoxide (DMSO) solution for cell culture applications Endotoxin tested SUMMARY Dimethylsulfoxide (DMSO) is a colorless organic solvent which enters the cell and distributes inside the cell to prevent the formation of damaging ice crystals during the freezing procedure CAS REGISTRY NO 67-68-5 ENDOTOXIN TESTED Yes LIQUID/POWDER Liquid PRODUCT CATEGORY Media supplements STERILE No STORAGE Room temperature (RT) MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80173 Supplier ILEX LIFE SCIENCES LLC Supplier No. P6036750100
PAN-Biotech Dimethyl Sulfoxide (DMSO) for Cell Culture Endotoxin Tested 100 ml DESCRIPTION PAN-Biotech Dimethyl Sulfoxide (DMSO) solution for cell culture applications Endotoxin tested SUMMARY Dimethylsulfoxide (DMSO) is a colorless organic solvent which enters the cell and distributes inside the cell to prevent the formation of damaging ice crystals during the freezing procedure CAS REGISTRY NO 67-68-5 ENDOTOXIN TESTED Yes LIQUID/POWDER Liquid PRODUCT CATEGORY Media supplements STERILE No STORAGE Room temperature (RT) MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany