Learn More
ILEX LIFE SCIENCES LLC PAN Biotech DPBS without Ca and Mg GMP Grade 500 ML 1/EA
SDP
Catalog No. 5000580357
Encompass_Preferred
PAN-Biotech DPBS without Ca and Mg GMP Grade 500 ml DESCRIPTION Dulbecco s Phosphate-Buffered Saline (DPBS) without calcium chloride and magnesium chloride GMP Grade Ready-to-use phosphate-buffered saline solution designed for direct use in GMP-regulated clinical and industrial applications It ensures reliable maintenance of pH and osmolality and provides essential inorganic ions required for safe and reproducible cell handling Combined with a very low endotoxin level high batch-to-batch consistency and comprehensive documentation DPBS GMP represents a reliable ready-to-use solution that can be immediately implemented in sensitive manufacturing and clinical workflows without additional preparation steps QUALITY GMP Grade (EU Annex 1) BICARBONATE Without Sodium Bicarbonate CALCIUM/MAGNESIUM Without Ca and Mg LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80357 Supplier ILEX LIFE SCIENCES LLC Supplier No. CT36500
Add to Cart
Add to Cart
PAN-Biotech DPBS without Ca and Mg GMP Grade 500 ml DESCRIPTION Dulbecco s Phosphate-Buffered Saline (DPBS) without calcium chloride and magnesium chloride GMP Grade Ready-to-use phosphate-buffered saline solution designed for direct use in GMP-regulated clinical and industrial applications It ensures reliable maintenance of pH and osmolality and provides essential inorganic ions required for safe and reproducible cell handling Combined with a very low endotoxin level high batch-to-batch consistency and comprehensive documentation DPBS GMP represents a reliable ready-to-use solution that can be immediately implemented in sensitive manufacturing and clinical workflows without additional preparation steps QUALITY GMP Grade (EU Annex 1) BICARBONATE Without Sodium Bicarbonate CALCIUM/MAGNESIUM Without Ca and Mg LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany