Learn More
ILEX LIFE SCIENCES LLC PAN Biotech EBSS Earles Balanced Salts Solution with Ca and Mg and phenol red without NaHCO3 500 ML 1/EA
SDP
Catalog No. 5000580178
Encompass_Preferred
PAN-Biotech Earle s Balanced Salts Solution (EBSS) w Ca and Mg w/o NaHCO3 500 ml INTRODUCTION Earle s Balanced Salts Solution (EBSS) with calcium and magnesium without sodium bicarbonate APPLICATION The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions These solutions are usually used for washing tissues and cells Often they are combined with other agents in cell and tissue treatment BICARBONATE without Sodium Bicarbonate CALCIUM/MAGNESIUM with Ca and Mg CONCENTRATION 1x LIQUID/POWDER liquid PHENOL RED with Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80178 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0430500
PAN-Biotech Earle s Balanced Salts Solution (EBSS) w Ca and Mg w/o NaHCO3 500 ml INTRODUCTION Earle s Balanced Salts Solution (EBSS) with calcium and magnesium without sodium bicarbonate APPLICATION The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions These solutions are usually used for washing tissues and cells Often they are combined with other agents in cell and tissue treatment BICARBONATE without Sodium Bicarbonate CALCIUM/MAGNESIUM with Ca and Mg CONCENTRATION 1x LIQUID/POWDER liquid PHENOL RED with Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures