Learn More
ILEX LIFE SCIENCES LLC PAN Biotech GBSS Geys Balanced Salt Solution with NaHCO3 cell suspension and washing solution 500 ML 1/EA
SDP
Catalog No. 5000580164
Encompass_Preferred
PAN-Biotech Gey s Balanced Salt Solution (GBSS) w 2 27 g/L NaHCO3 500 ml DESCRIPTION Gey s Balanced Salt Solution (GBSS) with calcium and magnesium with 2 27 g/L NaHCO3 APPLICATION GBSS is a phosphate bicarbonate buffered cell suspension and washing solution A complete balanced salt solution with a unique combination of salts GBSS contains the same five basic ions as EBSS HBSS and DPBS Among other applications GBSS has been used in organotypic mouse brain slice culture isolation of primary murine liver sinusoidal endothelial cells (LSECs) and organotypic hippocampal slice cultures BICARBONATE With 2 27 g/L Sodium Bicarbonate CALCIUM/MAGNESIUM With Ca and Mg CONCENTRATION 1x LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80164 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0448500
PAN-Biotech Gey s Balanced Salt Solution (GBSS) w 2 27 g/L NaHCO3 500 ml DESCRIPTION Gey s Balanced Salt Solution (GBSS) with calcium and magnesium with 2 27 g/L NaHCO3 APPLICATION GBSS is a phosphate bicarbonate buffered cell suspension and washing solution A complete balanced salt solution with a unique combination of salts GBSS contains the same five basic ions as EBSS HBSS and DPBS Among other applications GBSS has been used in organotypic mouse brain slice culture isolation of primary murine liver sinusoidal endothelial cells (LSECs) and organotypic hippocampal slice cultures BICARBONATE With 2 27 g/L Sodium Bicarbonate CALCIUM/MAGNESIUM With Ca and Mg CONCENTRATION 1x LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures