Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Glucose Solution for cell culture autoclaved 100 g per L 600 ML 1/EA
SDP
Catalog No. 5000580390
Encompass_Preferred
PAN-Biotech Glucose Solution for Cell Culture 100 g/L Autoclaved 6 x 100 ml DESCRIPTION PAN-Biotech Glucose Solution for Cell Culture 100 g/L Autoclaved Manufactured in Germany by PAN-Biotech LIQUID/POWDER liquid PRODUCT CATEGORY Media supplements PRODUCT TYPE Glucose Solution STERILE Yes STORAGE TEMP Room Temperature (RT) MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures This product may not be used as a pharmaceutical or veterinary drug agricultural product or food additive
Catalog No. 50-005-80390 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0401701X6
PAN-Biotech Glucose Solution for Cell Culture 100 g/L Autoclaved 6 x 100 ml DESCRIPTION PAN-Biotech Glucose Solution for Cell Culture 100 g/L Autoclaved Manufactured in Germany by PAN-Biotech LIQUID/POWDER liquid PRODUCT CATEGORY Media supplements PRODUCT TYPE Glucose Solution STERILE Yes STORAGE TEMP Room Temperature (RT) MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures This product may not be used as a pharmaceutical or veterinary drug agricultural product or food additive