Learn More
ILEX LIFE SCIENCES LLC PAN Biotech HBSS Hanks Balanced Salts Solution with Ca and Mg and NaHCO3 without phenol red 500 ML 1/EA
SDP
Catalog No. 5000580174
Encompass_Preferred
PAN-Biotech Hanks Balanced Salts Solution (HBSS) w/o Phenol red w Ca and Mg w 0 35 g/L NaHCO3 500 ml DESCRIPTION Hanks Balanced Salts Solution without Phenol red with calcium and magnesium with 0 35 g/L sodium bicarbonate APPLICATION HBSS can be used as a washing solution for living cells or as a carrier solution for various substances HBSS is used to maintain pH as well as osmotic balance and provides water and essential inorganic ions to cells It is designed for use with cells maintained in non-CO2 atmospheric conditions For cell dissociation procedures use a modified HBSS which is formulated without Ca and Mg salts BICARBONATE With 0 35 g/L Sodium Bicarbonate CALCIUM/MAGNESIUM With Ca and Mg CONCENTRATION 1x LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80174 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0432505
PAN-Biotech Hanks Balanced Salts Solution (HBSS) w/o Phenol red w Ca and Mg w 0 35 g/L NaHCO3 500 ml DESCRIPTION Hanks Balanced Salts Solution without Phenol red with calcium and magnesium with 0 35 g/L sodium bicarbonate APPLICATION HBSS can be used as a washing solution for living cells or as a carrier solution for various substances HBSS is used to maintain pH as well as osmotic balance and provides water and essential inorganic ions to cells It is designed for use with cells maintained in non-CO2 atmospheric conditions For cell dissociation procedures use a modified HBSS which is formulated without Ca and Mg salts BICARBONATE With 0 35 g/L Sodium Bicarbonate CALCIUM/MAGNESIUM With Ca and Mg CONCENTRATION 1x LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures