Learn More
ILEX LIFE SCIENCES LLC PAN Biotech HBSS Hanks Balanced Salts Solution without Ca and Mg and phenol red with NaHCO3 3000 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0434500X6
Encompass_Preferred
PAN-Biotech Hanks Balanced Salts Solution (HBSS) w/o Ca and Mg w/o Phenol red w 0 35 g/L NaHCO3 6 x 500 ml DESCRIPTION Hanks Balanced Salts Solution without Phenol red without calcium and magnesium with 0 35 g/L sodium bicarbonate APPLICATION HBSS can be used as a washing solution for living cells or as a carrier solution for various substances HBSS is used to maintain pH as well as osmotic balance and provides water and essential inorganic ions to cells It is designed for use with cells maintained in non-CO2 atmospheric conditions For cell dissociation procedures use a modified HBSS which is formulated without Ca and Mg salts BICARBONATE With 0 35 g/L Sodium Bicarbonate CALCIUM/MAGNESIUM Without Ca and Mg CONCENTRATION 1x LIQUID/POWDER Liquid PHENOL RED Without Phenol Red STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80346