Learn More
ILEX LIFE SCIENCES LLC PAN Biotech HEPES Buffer 1 M GMP Grade Cell Culture Buffer 100 ML 1/EA
SDP
Catalog No. 5000580307
Encompass_Preferred
PAN-Biotech HEPES Buffer 1 M GMP Grade 100 ml INTRODUCTION HEPES is a zwitterionic sulfonic acid buffering agent widely used in cell culture largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration (produced by aerobic respiration) when compared to bicarbonate buffers GMP GRADE PAN-Biotech HEPES Buffer 1 M GMP Grade is a ready-to-use highly concentrated buffer solution designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the proven HEPES formulation and is manufactured under strictly controlled GMP conditions Defined raw materials high batch-to-batch consistency and comprehensive documentation make HEPES Buffer 1 M GMP a true ready-to-use solution suitable for immediate use in sensitive cell culture and manufacturing workflows COMPOSITION HEPES 238 31 g/L QUALITY GMP Grade (EU Annex 1) LIQUID/POWDER Liquid STERILE Yes STORAGE 2-8 C
Catalog No. 50-005-80307 Supplier ILEX LIFE SCIENCES LLC Supplier No. CT01100
Add to Cart
Add to Cart
PAN-Biotech HEPES Buffer 1 M GMP Grade 100 ml INTRODUCTION HEPES is a zwitterionic sulfonic acid buffering agent widely used in cell culture largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration (produced by aerobic respiration) when compared to bicarbonate buffers GMP GRADE PAN-Biotech HEPES Buffer 1 M GMP Grade is a ready-to-use highly concentrated buffer solution designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the proven HEPES formulation and is manufactured under strictly controlled GMP conditions Defined raw materials high batch-to-batch consistency and comprehensive documentation make HEPES Buffer 1 M GMP a true ready-to-use solution suitable for immediate use in sensitive cell culture and manufacturing workflows COMPOSITION HEPES 238 31 g/L QUALITY GMP Grade (EU Annex 1) LIQUID/POWDER Liquid STERILE Yes STORAGE 2-8 C