Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Hygromycin B Solution 50 mg per ml cell culture selection antibiotic 120 ML 1/EA
SDP
Catalog No. 5000580235
Encompass_Preferred
PAN-Biotech Hygromycin B Solution 50 mg/ml 6 x 20 ml DESCRIPTION PAN-Biotech Hygromycin B Solution 50 mg/ml is an antibiotic commonly used in cell culture and laboratory research applications Hygromycin B is produced by the bacterium Streptomyces hygroscopicus It is an aminoglycoside that kills bacteria fungi and other eukaryotic cells by inhibiting protein synthesis APPLICATION Hygromycin B is used for the selection and maintenance of prokaryotic and eukaryotic cells that contain the hygromycin resistance gene The resistance gene is a kinase that inactivates hygromycin B through phosphorylation Since the discovery of hygromycin-resistance genes hygromycin B has become a standard selection antibiotic in gene transfer experiments in many prokaryotic and eukaryotic cells STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80235 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0608020X6
PAN-Biotech Hygromycin B Solution 50 mg/ml 6 x 20 ml DESCRIPTION PAN-Biotech Hygromycin B Solution 50 mg/ml is an antibiotic commonly used in cell culture and laboratory research applications Hygromycin B is produced by the bacterium Streptomyces hygroscopicus It is an aminoglycoside that kills bacteria fungi and other eukaryotic cells by inhibiting protein synthesis APPLICATION Hygromycin B is used for the selection and maintenance of prokaryotic and eukaryotic cells that contain the hygromycin resistance gene The resistance gene is a kinase that inactivates hygromycin B through phosphorylation Since the discovery of hygromycin-resistance genes hygromycin B has become a standard selection antibiotic in gene transfer experiments in many prokaryotic and eukaryotic cells STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures