Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Hygromycin B Solution 50 mg per ml cell culture selection antibiotic 20 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0608020
Encompass_Preferred
PAN-Biotech Hygromycin B Solution 50 mg/ml 20 ml DESCRIPTION PAN-Biotech Hygromycin B Solution 50 mg/ml is an antibiotic commonly used in cell culture and laboratory research applications Hygromycin B is produced by the bacterium Streptomyces hygroscopicus It is an aminoglycoside that kills bacteria fungi and other eukaryotic cells by inhibiting protein synthesis APPLICATION Hygromycin B is used for the selection and maintenance of prokaryotic and eukaryotic cells that contain the hygromycin resistance gene The resistance gene is a kinase that inactivates hygromycin B through phosphorylation Since the discovery of hygromycin-resistance genes hygromycin B has become a standard selection antibiotic in gene transfer experiments in many prokaryotic and eukaryotic cells STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80193