Learn More
ILEX LIFE SCIENCES LLC PAN Biotech IMDM Cell Culture Medium without glutamine with 25 mM HEPES with NaHCO3 500 ML 1/EA
SDP
Catalog No. 5000580191
Encompass_Preferred
PAN-Biotech IMDM Cell Culture Medium w/o L-Glutamine w 25 mM HEPES w 3 024 g/L NaHCO3 500 ml INTRODUCTION Iscove s Modified Dulbecco s Medium (IMDM) is a modification of Dulbecco s Modified Eagle Medium (DMEM) that includes selenium additional amino acids and vitamins sodium pyruvate and HEPES buffer In IMDM ferric nitrate (iron) is replaced with potassium nitrate IMDM is suitable for rapidly proliferating cultures of high cell density When supplemented with albumin transferrin and soy lipids it is highly suitable for culturing lymphocytes marrow cells or hybridoma cells BICARBONATE With 3 024 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE No Glutamine HEPES With 25 mM HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80191 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0420050
PAN-Biotech IMDM Cell Culture Medium w/o L-Glutamine w 25 mM HEPES w 3 024 g/L NaHCO3 500 ml INTRODUCTION Iscove s Modified Dulbecco s Medium (IMDM) is a modification of Dulbecco s Modified Eagle Medium (DMEM) that includes selenium additional amino acids and vitamins sodium pyruvate and HEPES buffer In IMDM ferric nitrate (iron) is replaced with potassium nitrate IMDM is suitable for rapidly proliferating cultures of high cell density When supplemented with albumin transferrin and soy lipids it is highly suitable for culturing lymphocytes marrow cells or hybridoma cells BICARBONATE With 3 024 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE No Glutamine HEPES With 25 mM HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures