Learn More
ILEX LIFE SCIENCES LLC PAN Biotech ITS Solution I 100x concentrated cell culture media supplement 10 ML 1/EA
SDP
Catalog No. 5000580166
Encompass_Preferred
PAN-Biotech ITS Solution I (100x conc ) Insulin-Transferrin-Selenium Cell Culture Media Supplement 10 ml DESCRIPTION PAN-Biotech ITS Solution I is a cell culture media supplement containing 0 5 mg/ml insulin human (recombinant) 0 5 mg/ml apo-Transferrin human and 0 5 g/ml sodium selenite in a balanced salt solution ITS solutions are commonly added to basal media to reduce the amount of fetal bovine serum (FBS) needed to culture cells or to promote optimal performance of serum-free media ITS Solutions can support routine maintenance and low density attachment of many adherent cell types in cultures with 2-4% FBS content CONCENTRATION 100x STERILE Yes STORAGE Store frozen at -20 C Once thawed keep solution at 2-8 C and avoid another freeze-thaw cycle COUNTRY OF ORIGIN Germany MANUFACTURER PAN-Biotech GmbH USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80166 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0703110
PAN-Biotech ITS Solution I (100x conc ) Insulin-Transferrin-Selenium Cell Culture Media Supplement 10 ml DESCRIPTION PAN-Biotech ITS Solution I is a cell culture media supplement containing 0 5 mg/ml insulin human (recombinant) 0 5 mg/ml apo-Transferrin human and 0 5 g/ml sodium selenite in a balanced salt solution ITS solutions are commonly added to basal media to reduce the amount of fetal bovine serum (FBS) needed to culture cells or to promote optimal performance of serum-free media ITS Solutions can support routine maintenance and low density attachment of many adherent cell types in cultures with 2-4% FBS content CONCENTRATION 100x STERILE Yes STORAGE Store frozen at -20 C Once thawed keep solution at 2-8 C and avoid another freeze-thaw cycle COUNTRY OF ORIGIN Germany MANUFACTURER PAN-Biotech GmbH USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures