Learn More
ILEX LIFE SCIENCES LLC PAN Biotech ITS Solution II 100x concentrated cell culture media supplement 5 ML 1/EA
SDP
Catalog No. 5000580165
Encompass_Preferred
PAN-Biotech ITS Solution II (100x conc ) Insulin-Transferrin-Selenium Cell Culture Media Supplement 5 ml DESCRIPTION Pan-Biotech ITS Solution II (100x) is a cell culture media supplement containing 1 mg/ml insulin human (recombinant) 0 55 mg/ml apo-Transferrin human and 0 5 g/ml sodium selenite in a balanced salt solution ITS solutions are commonly added to basal media to reduce the amount of fetal bovine serum (FBS) needed to culture cells or to promote optimal performance of serum-free media ITS Solutions can support routine maintenance and low density attachment of many adherent cell types in cultures with 2-4% FBS content LIQUID/POWDER Liquid CONCENTRATION 100x STERILE Yes STORAGE Store frozen at -20 C Once thawed keep solution at 2-8 C and avoid another freeze-thaw cycle COUNTRY OF ORIGIN Germany MANUFACTURER PAN-Biotech GmbH USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80165 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0703200
PAN-Biotech ITS Solution II (100x conc ) Insulin-Transferrin-Selenium Cell Culture Media Supplement 5 ml DESCRIPTION Pan-Biotech ITS Solution II (100x) is a cell culture media supplement containing 1 mg/ml insulin human (recombinant) 0 55 mg/ml apo-Transferrin human and 0 5 g/ml sodium selenite in a balanced salt solution ITS solutions are commonly added to basal media to reduce the amount of fetal bovine serum (FBS) needed to culture cells or to promote optimal performance of serum-free media ITS Solutions can support routine maintenance and low density attachment of many adherent cell types in cultures with 2-4% FBS content LIQUID/POWDER Liquid CONCENTRATION 100x STERILE Yes STORAGE Store frozen at -20 C Once thawed keep solution at 2-8 C and avoid another freeze-thaw cycle COUNTRY OF ORIGIN Germany MANUFACTURER PAN-Biotech GmbH USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures