Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Kanamycin Sulfate Solution 10 mg per ml cell culture antibiotic 100 ML 1/EA
SDP
Catalog No. 5000580183
Encompass_Preferred
PAN-Biotech Kanamycin Sulfate Solution 10 mg/ml for Cell Culture 100 ml DESCRIPTION PAN-Biotech Kanamycin Sulfate Solution 10 mg/ml is a broad-spectrum antibiotic purified from Streptomyces kanamyceticus bacterium it is used as a cell culture media supplement to control bacterial contamination It is effective effective against a wide variety of gram-negative and also some gram-positive bacteria APPLICATION Kanamycin Sulfate is often used as an antibiotic agent to prevent bacterial contamination of cell cultures COMPOSITION 10 000 00 g/L Kanamycin sulfate 9 000 00 g/L Sodium chloride LIQUID/POWDER liquid STERILE Yes STORAGE TEMPERATURE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80183 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0614100
PAN-Biotech Kanamycin Sulfate Solution 10 mg/ml for Cell Culture 100 ml DESCRIPTION PAN-Biotech Kanamycin Sulfate Solution 10 mg/ml is a broad-spectrum antibiotic purified from Streptomyces kanamyceticus bacterium it is used as a cell culture media supplement to control bacterial contamination It is effective effective against a wide variety of gram-negative and also some gram-positive bacteria APPLICATION Kanamycin Sulfate is often used as an antibiotic agent to prevent bacterial contamination of cell cultures COMPOSITION 10 000 00 g/L Kanamycin sulfate 9 000 00 g/L Sodium chloride LIQUID/POWDER liquid STERILE Yes STORAGE TEMPERATURE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures