Learn More
ILEX LIFE SCIENCES LLC PAN Biotech L Glutamine Penicillin Streptomycin Mix in saline solution cell culture media supplement 100 ML 1/EA
SDP
Catalog No. 5000580199
Encompass_Preferred
PAN-Biotech L-Glutamine Penicillin Streptomycin Solution (200 mM L-Glutamine 10 000 U/ml Penicillin 10 mg/ml Streptomycin in 0 9% NaCl) 100 ml DESCRIPTION Penicillin and streptomycin (Pen-Strep) antibiotics are used to prevent bacterial contamination of cell cultures given their combined action against gram-positive and gram-negative bacteria L-Glutamine an essential amino acid required by all mammalian and insect cells grown in culture serves as an additional source of carbon and energy and in particular is used for nucleic acid synthesis APPLICATION Used to supplement culture media to control bacterial contamination while also supplying energy to cells COMPOSITION 29 320 00 mg/L L-Glutamine 10 000 00 mg/L Streptomycin sulfate 9 000 00 mg/L Sodium chloride 10 000 000 00 Units/L Penicillin G potassium salt STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Lab Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80199 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0619100
PAN-Biotech L-Glutamine Penicillin Streptomycin Solution (200 mM L-Glutamine 10 000 U/ml Penicillin 10 mg/ml Streptomycin in 0 9% NaCl) 100 ml DESCRIPTION Penicillin and streptomycin (Pen-Strep) antibiotics are used to prevent bacterial contamination of cell cultures given their combined action against gram-positive and gram-negative bacteria L-Glutamine an essential amino acid required by all mammalian and insect cells grown in culture serves as an additional source of carbon and energy and in particular is used for nucleic acid synthesis APPLICATION Used to supplement culture media to control bacterial contamination while also supplying energy to cells COMPOSITION 29 320 00 mg/L L-Glutamine 10 000 00 mg/L Streptomycin sulfate 9 000 00 mg/L Sodium chloride 10 000 000 00 Units/L Penicillin G potassium salt STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Lab Research Use Only Not for use in diagnostic or therapeutic procedures