Learn More
ILEX LIFE SCIENCES LLC PAN Biotech MCDB 131 Cell Culture Medium with L Glutamine with NaHCO3 500 ML 1/EA
SDP
Catalog No. 5000580418
Encompass_Preferred
PAN-Biotech MCDB 131 Cell Culture Medium w L-Glutamine w 1 176 g/L NaHCO3 500 ml INTRO MCDB 131 was developed for cultivation of HMVEC under reduced serum content For this purpose it is often supplemented with serum EGF and hydrocortisone MCDB 131 is also suitable for other cell types (ex human omental microvascular cells hepatocytes myocytes smooth muscle cells hPSCs and MSCs) MCDB131 includes several components not found in traditional basal media such as trace elements putrescine adenine thymidine and higher levels of some amino acids and vitamins this allows it to be supplemented with very low levels of serum or defined components BICARBONATE With 1 176 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With L-Glutamine HEPES Without HEPES FORM Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only
Catalog No. 50-005-80418 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0480053
PAN-Biotech MCDB 131 Cell Culture Medium w L-Glutamine w 1 176 g/L NaHCO3 500 ml INTRO MCDB 131 was developed for cultivation of HMVEC under reduced serum content For this purpose it is often supplemented with serum EGF and hydrocortisone MCDB 131 is also suitable for other cell types (ex human omental microvascular cells hepatocytes myocytes smooth muscle cells hPSCs and MSCs) MCDB131 includes several components not found in traditional basal media such as trace elements putrescine adenine thymidine and higher levels of some amino acids and vitamins this allows it to be supplemented with very low levels of serum or defined components BICARBONATE With 1 176 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With L-Glutamine HEPES Without HEPES FORM Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only