Learn More
ILEX LIFE SCIENCES LLC PAN Biotech MCDB 153 Cell Culture Medium without Glutamine with NaHCO3 500 ML 1/EA
SDP
Catalog No. 5000580195
Encompass_Preferred
PAN-Biotech MCDB 153 Cell Culture Medium w/o L-Glutamine w 1 176 g/L NaHCO3 500 ml INTRODUCTION MCDB media were designed for serum-free growth of specific cell types Each MCDB medium was formulated to provide a defined and optimally balanced nutritional environment that selectively promotes growth of a specific cell type MCDB 153 medium is a modification of Ham s F12 Medium designed for the growth of human keratinocytes chicken embryo fibroblasts and Chinese Hamster Ovary (CHO) cells BICARBONATE With 1 176 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE No Glutamine HEPES With HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80195 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0480062
PAN-Biotech MCDB 153 Cell Culture Medium w/o L-Glutamine w 1 176 g/L NaHCO3 500 ml INTRODUCTION MCDB media were designed for serum-free growth of specific cell types Each MCDB medium was formulated to provide a defined and optimally balanced nutritional environment that selectively promotes growth of a specific cell type MCDB 153 medium is a modification of Ham s F12 Medium designed for the growth of human keratinocytes chicken embryo fibroblasts and Chinese Hamster Ovary (CHO) cells BICARBONATE With 1 176 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE No Glutamine HEPES With HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures