Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Medium 199 M199 Cell Culture Medium with EBSS and L Glutamine GMP Grade 500 ML 1/EA
SDP
Catalog No. 5000580356
Encompass_Preferred
PAN-Biotech Medium 199 (M199) w EBSS w L-Glutamine w 2 2 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION Medium 199 (M199) GMP Grade is a ready-to-use classical basal cell culture medium designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the well-established M199 formulation and is manufactured under strictly controlled GMP conditions Defined raw materials high batch-to-batch consistency and comprehensive documentation make M199 GMP a true ready-to-use solution suitable for immediate use in development scale-up and manufacturing workflows EBSS Contains Earle s Balanced Salt Solution QUALITY GMP Grade (EU Annex 1) BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With L-Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80356 Supplier ILEX LIFE SCIENCES LLC Supplier No. CT07050
PAN-Biotech Medium 199 (M199) w EBSS w L-Glutamine w 2 2 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION Medium 199 (M199) GMP Grade is a ready-to-use classical basal cell culture medium designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the well-established M199 formulation and is manufactured under strictly controlled GMP conditions Defined raw materials high batch-to-batch consistency and comprehensive documentation make M199 GMP a true ready-to-use solution suitable for immediate use in development scale-up and manufacturing workflows EBSS Contains Earle s Balanced Salt Solution QUALITY GMP Grade (EU Annex 1) BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With L-Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany