Learn More
ILEX LIFE SCIENCES LLC PAN Biotech MEM Eagle Cell Culture Medium with EBSS with Stable Glutamine with 25 mM HEPES 500 ML 1/EA
SDP
Catalog No. 5000580176
Encompass_Preferred
PAN-Biotech MEM Eagle Cell Culture Medium w EBSS w Stable Glutamine w 25 mM HEPES w 2 2 g/L NaHCO3 500 ml INTRODUCTION Minimum Essential Medium (MEM) is a widely used synthetic cell culture media After finding that Eagle s Basal Medium (BME) did not meet specific nutritional requirements of normal mammalian fibroblasts and certain HeLa cells MEM was created with higher concentrations of amino acids to more closely resemble the protein composition of mammalian cells When supplemented with serum MEM is suitable for cultivation of a wide variety of cells grown in monolayers BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With Stable Glutamine HEPES With 25 mM HEPES LIQUID/POWEDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80176 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0408250
PAN-Biotech MEM Eagle Cell Culture Medium w EBSS w Stable Glutamine w 25 mM HEPES w 2 2 g/L NaHCO3 500 ml INTRODUCTION Minimum Essential Medium (MEM) is a widely used synthetic cell culture media After finding that Eagle s Basal Medium (BME) did not meet specific nutritional requirements of normal mammalian fibroblasts and certain HeLa cells MEM was created with higher concentrations of amino acids to more closely resemble the protein composition of mammalian cells When supplemented with serum MEM is suitable for cultivation of a wide variety of cells grown in monolayers BICARBONATE With 2 2 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With Stable Glutamine HEPES With 25 mM HEPES LIQUID/POWEDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures