Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Pancoll Mouse Cell Separating Solution 1 086 g per ml density 100 ML 1/EA
SDP
Catalog No. 5000580171
Encompass_Preferred
Pancoll Mouse Cell Separating Solution (Centrifugation Media) Density 1 086 g/ml 100 ml INTRODUCTION Pancoll separation media (density gradient media) contain a polysaccharide with a molecular weight of 400 000 daltons This hydrophilic polymer allows production of aqueous solutions for cell separation with a density of up to 1 119 g/ml PAN-Biotech offers a variety of ready-to-use products with a density of 1 063 g/ml up to 1 119 g/ml for a very wide range of cell isolation applications SPECIES Optimized for mouse samples DENSITY 1 086 g/mL EDTA Without EDTA HEPES Without HEPES LIQUID/POWDER Liquid STERILE Yes STORAGE 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany COMPOSITION 97 000 00 mg/L Polysucrose 78 000 00 mg/L Diatrizoic Acid USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80171 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0464100
Pancoll Mouse Cell Separating Solution (Centrifugation Media) Density 1 086 g/ml 100 ml INTRODUCTION Pancoll separation media (density gradient media) contain a polysaccharide with a molecular weight of 400 000 daltons This hydrophilic polymer allows production of aqueous solutions for cell separation with a density of up to 1 119 g/ml PAN-Biotech offers a variety of ready-to-use products with a density of 1 063 g/ml up to 1 119 g/ml for a very wide range of cell isolation applications SPECIES Optimized for mouse samples DENSITY 1 086 g/mL EDTA Without EDTA HEPES Without HEPES LIQUID/POWDER Liquid STERILE Yes STORAGE 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany COMPOSITION 97 000 00 mg/L Polysucrose 78 000 00 mg/L Diatrizoic Acid USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures