Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Paneticin G418 50 mg per ml cell culture selection antibiotic 20 ML 1/EA
SDP
Catalog No. 5000580162
Encompass_Preferred
PAN-Biotech Paneticin G418 (G418 Sulfate) 50 mg/ml Selective Antibiotic for Cell Culture 20 ml DESCRIPTION Paneticin G418 (G418 disulfate salt) is an aminoglycoside antibiotic similar in structure to gentamicin that is used for selection and maintenance of prokaryotic and eukaryotic cells transfected with an iNOS promoter construct and neomycin resistance gene It has a broad range of antibacterial activity and activity against protozoa cecal amoebiasis and helminths Optimal concentrations for selection and maintenance vary by cell lines (between 100 - 500 g/ml) Paneticin G418 is a suitable alternative to Geneticin Selective Antibiotic LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80162 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0616220
PAN-Biotech Paneticin G418 (G418 Sulfate) 50 mg/ml Selective Antibiotic for Cell Culture 20 ml DESCRIPTION Paneticin G418 (G418 disulfate salt) is an aminoglycoside antibiotic similar in structure to gentamicin that is used for selection and maintenance of prokaryotic and eukaryotic cells transfected with an iNOS promoter construct and neomycin resistance gene It has a broad range of antibacterial activity and activity against protozoa cecal amoebiasis and helminths Optimal concentrations for selection and maintenance vary by cell lines (between 100 - 500 g/ml) Paneticin G418 is a suitable alternative to Geneticin Selective Antibiotic LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures