Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Panexin CD Serum Substitute Chemically Defined Animal-Free 50 ML 1/EA
SDP
Catalog No. 5000580200
Encompass_Preferred
PAN-Biotech Panexin CD Serum Replacement with Chemically-Defined Components 50 ml DESCRIPTION Panexin CD is a chemically defined xeno-free serum substitute for cultivation of adherent or non-adherent cells under serum-free or serum-reduced culture conditions It supports the growth of many cell types in an optimum manner without any extra handling compared to fetal bovine serum (FBS) or human serum COMPOSITION Panexin CD is free of any animal or human-derived components It contains recombinant proteins lipids salts amino acids trace elements hormones and a 3-dimensional substrate release system in an optimized formulation It contains no growth factors undefined hydrolysates or peptones CELL TYPE Various CULTURE TYPE Adherent culture or suspension culture LIQUID/POWDER liquid STERILE Yes STORAGE -20 C in the dark MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80200 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0493050
PAN-Biotech Panexin CD Serum Replacement with Chemically-Defined Components 50 ml DESCRIPTION Panexin CD is a chemically defined xeno-free serum substitute for cultivation of adherent or non-adherent cells under serum-free or serum-reduced culture conditions It supports the growth of many cell types in an optimum manner without any extra handling compared to fetal bovine serum (FBS) or human serum COMPOSITION Panexin CD is free of any animal or human-derived components It contains recombinant proteins lipids salts amino acids trace elements hormones and a 3-dimensional substrate release system in an optimized formulation It contains no growth factors undefined hydrolysates or peptones CELL TYPE Various CULTURE TYPE Adherent culture or suspension culture LIQUID/POWDER liquid STERILE Yes STORAGE -20 C in the dark MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures