Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Penicillin Streptomycin Amphotericin B Mix in saline solution cell culture antimicrobial 125 ML 1/EA
SDP
Catalog No. 5000580192
Encompass_Preferred
PAN-Biotech Penicillin-Streptomycin-Amphotericin B Mix (10 000 U/ml Penicillin 10 mg/ml Streptomycin 25 g/ml Amphotericin B) in 0 85% Saline Solution 25 x 5 ml DESCRIPTION Penicillin and streptomycin antibiotics (together known as Pen-Strep) are used to prevent bacterial contamination of cell cultures to due their effective combined action against gram-positive and gram-negative bacteria Amphotericin B is an antifungal agent that protects cell cultures from contamination by yeast and multicellular fungi APPLICATION Penicillin-Streptomycin-Amphotericin B Mix is used to supplement cell culture media to control both bacterial and fungal contamination COMPONENTS Amphotericin B 25 00 mg/L Streptomycin sulfate 10 000 00 mg/L Sodium chloride 8 500 00 mg/L Penicillin G potassium salt 10 000 000 00 Units/L STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80192 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0607325
PAN-Biotech Penicillin-Streptomycin-Amphotericin B Mix (10 000 U/ml Penicillin 10 mg/ml Streptomycin 25 g/ml Amphotericin B) in 0 85% Saline Solution 25 x 5 ml DESCRIPTION Penicillin and streptomycin antibiotics (together known as Pen-Strep) are used to prevent bacterial contamination of cell cultures to due their effective combined action against gram-positive and gram-negative bacteria Amphotericin B is an antifungal agent that protects cell cultures from contamination by yeast and multicellular fungi APPLICATION Penicillin-Streptomycin-Amphotericin B Mix is used to supplement cell culture media to control both bacterial and fungal contamination COMPONENTS Amphotericin B 25 00 mg/L Streptomycin sulfate 10 000 00 mg/L Sodium chloride 8 500 00 mg/L Penicillin G potassium salt 10 000 000 00 Units/L STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures