Learn More
ILEX LIFE SCIENCES LLC PAN Biotech PHA L Phytohemagglutinin L from Phaseolus vulgaris Lyophilized 2 MG 1/EA
SDP
Catalog No. 5000580253
Encompass_Preferred
PAN-Biotech Phytohemagglutinin-L (PHA-L) from Phaseolus vulgaris Lyophilized 2 mg DESCRIPTION Phytohemagglutinin-L (PHA-L) is a plant lectin purified from Phaseolus vulgaris (red kidney bean) Lectins are an important mitogen group Lectins bind specifically to membrane-glycoproteins of different cells including lymphocytes The agglutinating effect leads to lymphocyte activation and proliferation APPLICATION Plant lectin suitable for stimulating lymphocyte proliferation and differentiation CAS NO 9008-97-3 LIQUID/POWDER Powder STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany OTHER NAMES Phaseolus Vulgaris Lectin L PHA L subunit Leucoagglutinating phytohemagglutinin DLEC2 Leucoagglutinin UniProtKB P05087 USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80253 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0501011M
PAN-Biotech Phytohemagglutinin-L (PHA-L) from Phaseolus vulgaris Lyophilized 2 mg DESCRIPTION Phytohemagglutinin-L (PHA-L) is a plant lectin purified from Phaseolus vulgaris (red kidney bean) Lectins are an important mitogen group Lectins bind specifically to membrane-glycoproteins of different cells including lymphocytes The agglutinating effect leads to lymphocyte activation and proliferation APPLICATION Plant lectin suitable for stimulating lymphocyte proliferation and differentiation CAS NO 9008-97-3 LIQUID/POWDER Powder STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany OTHER NAMES Phaseolus Vulgaris Lectin L PHA L subunit Leucoagglutinating phytohemagglutinin DLEC2 Leucoagglutinin UniProtKB P05087 USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures