Learn More
ILEX LIFE SCIENCES LLC PAN Biotech RPMI 1640 Cell Culture Medium without glutamine without glucose with 2 g per L NaHCO3 3000 ML 1/EA
SDP
Catalog No. 5000580223
Encompass_Preferred
PAN-Biotech RPMI 1640 Cell Culture Medium w/o L-Glutamine w/o Glucose w 2 0 g/L NaHCO3 6 x 500 ml INTRODUCTION RPMI 1640 also known as RPMI Medium was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells We offer a variety of RPMI media to meet the needs of your specific applications BICARBONATE With 2 0 g/L Sodium Bicarbonate GLUCOSE Without glucose GLUTAMINE No Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80223 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0417550X6
PAN-Biotech RPMI 1640 Cell Culture Medium w/o L-Glutamine w/o Glucose w 2 0 g/L NaHCO3 6 x 500 ml INTRODUCTION RPMI 1640 also known as RPMI Medium was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells We offer a variety of RPMI media to meet the needs of your specific applications BICARBONATE With 2 0 g/L Sodium Bicarbonate GLUCOSE Without glucose GLUTAMINE No Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED With Phenol Red SODIUM PYRUVATE Without Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures