Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Sodium Bicarbonate Solution 75000 mg NaHCO3 per L cell culture media supplement 100 ML 1/EA
SDP
Catalog No. 5000580179
Encompass_Preferred
PAN-Biotech Sodium Bicarbonate 7 5% Solution (NaHCO3) cell culture media supplement 100 ml DESCRIPTION PAN-Biotech Sodium Bicarbonate 7 5% Solution (NaHCO3) is an alkaline agent commonly used as a cell culture media supplement for the maintenance of appropriate pH when cells are incubated with carbon dioxide Additionally sodium bicarbonate provides some nutritional value when culturing mammalian cells COMPOSITION 75 000 00 mg/L NaHCO CAS NO 144-55-8 LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80179 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0444100
PAN-Biotech Sodium Bicarbonate 7 5% Solution (NaHCO3) cell culture media supplement 100 ml DESCRIPTION PAN-Biotech Sodium Bicarbonate 7 5% Solution (NaHCO3) is an alkaline agent commonly used as a cell culture media supplement for the maintenance of appropriate pH when cells are incubated with carbon dioxide Additionally sodium bicarbonate provides some nutritional value when culturing mammalian cells COMPOSITION 75 000 00 mg/L NaHCO CAS NO 144-55-8 LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures