Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Sodium Pyruvate 100 mM Solution cell culture media supplement 100 ML 1/EA
SDP
Catalog No. 5000580180
Encompass_Preferred
PAN-Biotech Sodium Pyruvate Solution 100 mM cell culture media supplement 100 ml DESCRIPTION Sodium Pyruvate Solution finds application in cell culture as an easily accessible carbohydrate source for cells Sodium pyruvate is also involved with amino acid metabolism and initiates the Kreb s cycle In general the 100 mM solution should be diluted 1 100 for most cell culture applications COMPOSITION 11 050 00 mg/L Sodium Pyruvate CAS NO 113-24-6 LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80180 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0443100
PAN-Biotech Sodium Pyruvate Solution 100 mM cell culture media supplement 100 ml DESCRIPTION Sodium Pyruvate Solution finds application in cell culture as an easily accessible carbohydrate source for cells Sodium pyruvate is also involved with amino acid metabolism and initiates the Kreb s cycle In general the 100 mM solution should be diluted 1 100 for most cell culture applications COMPOSITION 11 050 00 mg/L Sodium Pyruvate CAS NO 113-24-6 LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures