Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Trypsin EDTA in DPBS Solution without Ca and Mg GMP Grade 100 ML 1/EA
Supplier: ILEX LIFE SCIENCES LLC CT023100
PAN-Biotech Trypsin 0 05% / EDTA 0 02% in DPBS w/o Ca and Mg GMP Grade 100 ml DESCRIPTION Trypsin 0 05% / EDTA 0 02% GMP Grade (EU Annex 1) is a porcine pancreas-derived enzyme plus EDTA in a DPBS solution This ready-to-use enzymatic cell detachment solution designed for direct use in GMP-regulated clinical and industrial applications It enables gentle and reproducible dissociation of adherent cells and tissues supporting controlled and robust cell processing workflows The moderate trypsin concentration makes this formulation particularly suitable for sensitive cell lines and controlled detachment procedures as required in clinical cell processing cell and gene therapy and biopharmaceutical manufacturing Combined with high batch-to-batch consistency defined quality parameters and comprehensive documentation this product represents a true ready-to-use solution suitable for immediate implementation in sensitive manufacturing and clinical processes