Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Trypsin Inhibitor III 1 mg per ml solution animal and human free 50 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P10034100
Encompass_Preferred
PAN-Biotech Trypsin Inhibitor III 1 mg/ml in PBS free of animal human components 50 ml DESCRIPTION Trypsin Inhibitor III is a sterile solution of purified proteins to inhibit trypsin proteolytic activity It does not contain any animal or human components Supplied as 1 mg/ml in PBS solution COMPOSITION Trypsin Inhibitor III solution contains natural protease inhibitor proteins The biological activity is depending on trypsin concentration APPLICATION For inhibiting trypsin activity in serum-free culture of adherent cells Trypsin is used for dissociation of adhesion-dependent cells Serum contains natural trypsin inhibitors In serum-free cell culture the neutralizing effect of serum is absent Therefore it is crucial to completely block proteolytic activity from trypsin after detachment of the cells Inhibits trypsin activity chymotrypsin and other proteolytic enzymes are inhibited to a lesser extent
Catalog No. 50-005-80169