Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Trypsin Inhibitor Solution 1 mg per ml sterile 100 ML 1/EA
SDP
Catalog No. 5000580186
Encompass_Preferred
PAN-Biotech Trypsin Inhibitor Solution 1 mg/ml 100 ml DESCRIPTION A sterile solution of plant-based inhibitors against trypsin and in some effect to chymotrypsin and plasmin does not inhibit metalloproteases tissue based kallikrein and acid or thioproteases APPLICATION Inhibiting trypsin activity in serum-free culture of adherent cells In serum-free culture the natural neutralizing effect of serum trypsin inhibitors is absent thus it is crucial to completely block proteolytic activity from trypsin after detachment of the cells ADVANTAGE Trypsin Inhibitor Solution contains protease inhibitors as well as growth promoting agents After detachment and re-suspension in serum-free medium Trypsin Inhibitor Solution supports the recovery attachment and spreading of the cells The usual centrifugation step to remove trypsin inhibitor from the cells should be omitted with this product This time-saving feature facilitates a more convenient efficient serum-free cell culture
Catalog No. 50-005-80186 Supplier ILEX LIFE SCIENCES LLC Supplier No. P10033100
PAN-Biotech Trypsin Inhibitor Solution 1 mg/ml 100 ml DESCRIPTION A sterile solution of plant-based inhibitors against trypsin and in some effect to chymotrypsin and plasmin does not inhibit metalloproteases tissue based kallikrein and acid or thioproteases APPLICATION Inhibiting trypsin activity in serum-free culture of adherent cells In serum-free culture the natural neutralizing effect of serum trypsin inhibitors is absent thus it is crucial to completely block proteolytic activity from trypsin after detachment of the cells ADVANTAGE Trypsin Inhibitor Solution contains protease inhibitors as well as growth promoting agents After detachment and re-suspension in serum-free medium Trypsin Inhibitor Solution supports the recovery attachment and spreading of the cells The usual centrifugation step to remove trypsin inhibitor from the cells should be omitted with this product This time-saving feature facilitates a more convenient efficient serum-free cell culture