Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Williams Medium E without glucose without glutamine without phenol red 500 ML 1/EA
SDP
Catalog No. 5000580226
Encompass_Preferred
PAN-Biotech William s Medium E w/o Glucose w/o L-Glutamine w/o Phenol red w 2 24 g/L NaHCO3 500 ml INTRODUCTION Williams and Gunn originally developed William s E Medium as a reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells William s E Medium may also be used for the cultivation of primary hepatocyte cells of other species as well as human-derived HepaRG cells The medium is unique in that it contains zinc iron manganese non-essential amino acids the reducing agent glutathione and the lipid methyl linoleate BICARBONATE With 2 24 g/L Sodium Bicarbonate GLUCOSE Without glucose GLUTAMINE No Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED Without Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80226 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0429050S4
PAN-Biotech William s Medium E w/o Glucose w/o L-Glutamine w/o Phenol red w 2 24 g/L NaHCO3 500 ml INTRODUCTION Williams and Gunn originally developed William s E Medium as a reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells William s E Medium may also be used for the cultivation of primary hepatocyte cells of other species as well as human-derived HepaRG cells The medium is unique in that it contains zinc iron manganese non-essential amino acids the reducing agent glutathione and the lipid methyl linoleate BICARBONATE With 2 24 g/L Sodium Bicarbonate GLUCOSE Without glucose GLUTAMINE No Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED Without Phenol Red SODIUM PYRUVATE With Sodium Pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures