Learn More
Sino Biological HCoV-OC43 Spike Antibody, Mouse MAb
Supplier:  Sino Biological 68086MM29100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant CoV spike S1+S2 ECD protein. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-222-1623