Learn More
Biocare Medical, LLC Pan Cytokeratin [Lu-5] Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM043AA
Encompass_Preferred
Pan Cytokeratin [Lu-5] antibody is has been demonstrated as a useful marker for the differentiation of epithelial and mesothelial cells from mesenchymal cells in normal and tumor tissues. It has been shown to serve as a first-order pan cytokeratin antibody for both acidic (type I) and basic (type II) cytokeratin subfamilies of all vertebrates tested so far. In immunohistochemical studies, [Lu-5] stains an intracytoplasmic, formaldehyde-resistant epitope on the surface of cytokeratin filaments. [Lu-5] has been shown to be superior to [AE1/AE3].
Catalog No. 50-830-15