Learn More
Abcam pan-HMR GSK3 beta ELISA Kit, 1 x 96 Tests
Catalog No. 502698629
Encompass_Preferred
pan-HMR GSK3 beta ELISA Kit is a single-wash 90-min Simplestep used to quantify pan-HMR GSK3 beta with a sensitivity of 34.77 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8629 Supplier Abcam Supplier No. AB3150671X96TESTS
pan-HMR GSK3 beta ELISA Kit is a single-wash 90-min Simplestep used to quantify pan-HMR GSK3 beta with a sensitivity of 34.77 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement