Learn More
Abcam pan-HMR VEGFC ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3150501X96TESTS
Encompass_Preferred
pan-HMR VEGFC ELISA Kit is a single-wash 90-min Simplestep used to quantify pan-HMR VEGFC with a sensitivity of 4.375 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2738511