Learn More
Thomas Scientific Lab Ice Pan w/lid, Magic TouchT 2, Midi, black, 4L, expanded urethane, lightweight, chemical-resistant, snug lid, pour spout, fill line indicator, raised top edge, easy transport, textured exterior, no-slip finish, 11.95 x9.01 x8.57"H w/lid
SDP
Catalog No. 501460135
Encompass_Preferred
Superior insulating properties combined with high chemical resistance make Magic Touch 2™ Ice Pans compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen. Their lightly textured non-slip finish and raised edge provides secure transport and a stylish look. Magic Touch 2™ Ice Pans are available in 1.0L Mini, 4.0L Midi and 9.0L Maxi sizes, each in four colors; ebony, cherry red, true blue and emerald green. Expanded urethane laboratory ice pans are lightweight, durable and non-sweating Impervious to moisture and odors Able to withstand a wide temperature range: -196ÝC (-320ÝF) to 100ÝC (212ÝF) Features convenient pour spout, drip-proof rims and fill line indicator* Snug-fitting, keyed lid and base make stacking easy and secure Note: 1L pan does not include pour spout or fill line indicator
This item is not returnable. View return policy
Catalog No. 50-146-0135 Supplier Thomas Scientific Supplier No. 1209Q84
Superior insulating properties combined with high chemical resistance make Magic Touch 2™ Ice Pans compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen. Their lightly textured non-slip finish and raised edge provides secure transport and a stylish look. Magic Touch 2™ Ice Pans are available in 1.0L Mini, 4.0L Midi and 9.0L Maxi sizes, each in four colors; ebony, cherry red, true blue and emerald green. Expanded urethane laboratory ice pans are lightweight, durable and non-sweating Impervious to moisture and odors Able to withstand a wide temperature range: -196ÝC (-320ÝF) to 100ÝC (212ÝF) Features convenient pour spout, drip-proof rims and fill line indicator* Snug-fitting, keyed lid and base make stacking easy and secure Note: 1L pan does not include pour spout or fill line indicator