Learn More
Hach Company PAN Indicator Solution, 0.1%, 100 mL
Supplier:  Hach Company 2122432
Encompass
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn. 100 mL bottle
Catalog No. 50-199-5453