Learn More
Hach Company PAN Indicator Solution, 0.1%, 50 mL SCDB
Supplier:  Hach Company 2122426
Encompass
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn. 50 mL Self-Contained Dropping Bottle.
Catalog No. 50-199-5452