Learn More
Hach Company PAN Indicator Solution, 0.3%, 100 mL
Supplier:  Hach Company 2150232
Encompass
For low-level determination of nickel and cobalt by the PAN method. Hach method 8078. 100 mL bottle
Catalog No. 50-199-5474