Learn More
Sino Biological Pan Influenza A Nucleoprotein Antibody, Rabbit MAb 100ug
Supplier:  Sino Biological 40208R117100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP Protein (Catalog# 40208-V08B; P22435.2; Met1-Asn498).
Catalog No. 50-224-7432