Learn More
Sino Biological Pan Influenza A Nucleoprotein Antibody (HRP), Rabbit MAb 100 µg
Catalog No. 5000704863
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP Protein (Catalog# 40208-V08B; P22435.2; Met1-Asn498), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-007-04863 Supplier Sino Biological Supplier No. 40208R014H100
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP Protein (Catalog# 40208-V08B; P22435.2; Met1-Asn498), and conjugated with horseradish-peroxidase (HRP).