Learn More
Sino Biological Pan Influenza B Nucleoprotein Antibody, Mouse MAb 100ug
Supplier:  Sino Biological 40438MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza B (B/Florida/4/2006) Nucleoprotein / NP Protein (Catalog# 40438-V08B; ACF54251.1; Met1-Tyr560). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7367