Learn More
Medchemexpress LLC Pan-KRAS-IN-18 | 2937327-83-6 | 99.9% | 450.56 | 5 MG
SDP
Catalog No. 5000391613
Encompass_Preferred
pan-KRAS-IN-18 is a panKRAS inhibitor that inhibits KRAS WT and KRAS G12V with IC50s of 29 and 9 nM, respectively. It exhibits antiproliferative activity in KRAS-mutant cell lines and can be used for lung cancer research.
  • Inhibits K-Ras WT (29 nM IC50) and KRas G12V (9 nM IC50)
  • Exhibits antiproliferative effects against Ba/F3 KRASG12V and NCI-H727 cells with EC50s of 0.959 and >1 μM, respectively (72 h)
  • Shows high clearance in mouse and human liver microsomes
  • Displays high binding affinity towards KRASG12V with a Ki of 25 nM
  • Appearance is solid, white to off-white
Catalog No. 50-003-91613 Supplier Medchemexpress LLC Supplier No. HY1629605MG
pan-KRAS-IN-18 is a panKRAS inhibitor that inhibits KRAS WT and KRAS G12V with IC50s of 29 and 9 nM, respectively. It exhibits antiproliferative activity in KRAS-mutant cell lines and can be used for lung cancer research.
  • Inhibits K-Ras WT (29 nM IC50) and KRas G12V (9 nM IC50)
  • Exhibits antiproliferative effects against Ba/F3 KRASG12V and NCI-H727 cells with EC50s of 0.959 and >1 μM, respectively (72 h)
  • Shows high clearance in mouse and human liver microsomes
  • Displays high binding affinity towards KRASG12V with a Ki of 25 nM
  • Appearance is solid, white to off-white