Learn More
Selleck Chemical LLC LY3009120 5mg 1454682-72-4
SDP
Supplier:  Selleck Chemical LLC S78425MG
Encompass_Preferred
LY03009120 is a potent pan-Raf inhibitor with IC50 of 44 nM, 31-47 nM, and 42 nM for A-raf, B-Raf, and C-Raf in A375 cells, respectively. LY03009120 induces autophagy. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1038738