Learn More
Medchemexpress LLC HY-101295 10mg Medchemexpress, Pan-RAS-IN-1 CAS:1835283-94-7 Purity:>98%
SDP
Catalog No. 502028659
Encompass_Preferred
Medchemexpress, HY-101295 10mg Pan-RAS-IN-1 CAS:1835283-94-7 Pan-RAS-IN-1 is a pan-Ras inhibitor that disrupts the interaction of Ras proteins and their effectors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8659 Supplier Medchemexpress LLC Supplier No. HY10129510MG
Medchemexpress, HY-101295 10mg Pan-RAS-IN-1 CAS:1835283-94-7 Pan-RAS-IN-1 is a pan-Ras inhibitor that disrupts the interaction of Ras proteins and their effectors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.