Learn More
TARGETMOL CHEMICALS INC PANADIPLON 5MG
SDP
Catalog No. 502580525
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Panadiplon (FG 10571) is a selective gamma-aminobutyric acid receptor agonist and partial agonist of 5GABAA a benzodiazepine receptor used in the treatment of anxiety disorders.Panadiplon exhibits selectivity for 5GABAA receptors versus 1GABAA receptors. purity: 99%
Catalog No. 50-258-0525 Supplier TARGETMOL CHEMICALS INC Supplier No. T282945MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Panadiplon (FG 10571) is a selective gamma-aminobutyric acid receptor agonist and partial agonist of 5GABAA a benzodiazepine receptor used in the treatment of anxiety disorders.Panadiplon exhibits selectivity for 5GABAA receptors versus 1GABAA receptors. purity: 99%